2-Deoxy-D(-)-ribose 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:533-67-5
:
Formula:C₅H₁₀O₄
MDL Number:MFCD00135904
:134.13 g/mol
Merck Index:13,02925
Boiling Pt:292 °C (1013 hPa)
Melting Pt:91 °C
Storage Temperature:Ambient
EINECS:208-573-0
Specification Test Results
- Appearance:White to light yellow Crystalline powder
- Infrared spectrum:Conforms
- Assay:≥98.5 %
- Loss on drying:≤1.0 % (40°C, 2 hrs) (vacuum)
- Residue after ignition:≤0.5 %
- Specific optical rotation:-55° to -59° (20°C, 589 nm) (c=1, H2O, 24 hrs.)
- Heavy metals (as Pb):≤20 ppm