Order Entry
Sweden
Orders LinkContactUsLinkComponent
Acetone

Acetone

Flammable
Irritant
Supplier:   ABSOR
Acetone
Supplier:  ABSOR
Certificates
Acetone

2 Options Available Below

Product Details & Documents
Synonyms:  2-Propanone, DMK, Propanone, Dimethyl ketone
  • CAS Number:67-64-1
  • Formula:CH₃COCH₃
  • MDL Number:MFCD00008765
  • Molecular Weight:58.08 g/mol
  • Merck Index:13,00067
  • Boiling Pt:56.2 °C (1013 hPa)
  • Melting Pt:–95.4 °C
  • Density:0.792 g/cm³ (20 °C)
  • Flash Pt:<–20 °C (closed cup)
  • Storage Temperature:Ambient
  • EINECS:200-662-2
  • UN:1090
  • ADR:3,II
  • REACH:01-2119471330-49
Product Family Options

Product Information

  • Pk
    Availability
    Price
  • 5 L
    Log in for availability
    Log in for availability
    1 * 5 L
    1 * 5 L - 1 225,00 kr
    1 225,00 kr
    1 * 5 L
       1 225,00 kr

  • 10 L
    Log in for availability
    Log in for availability
    1 * 10 L
    1 * 10 L - 2 250,00 kr
    2 250,00 kr
    1 * 10 L
       2 250,00 kr