Order Entry
Sweden
ContactUsLinkComponent
Electrical energy sources
Electrical energy sources
Catalog # 765-0109
undefined
Electrical energy sources
Catalog # 765-0109
Specifications
  • Description:
    Electrical energy sources, experimental kit
  • Pk:
    1
Specifications
About this item
The kit contains about 15 different components with which a number of experiments can be performed. The components are designed so that students can quickly and easily perform their laboratory work. The kit can show how the following forms of energy are generated:
  • Thermal energy (thermocouple)
  • Radiant energy (solar cell)
  • Chemical energy (galvanic element)
  • Mechanical energy (induction)
To demonstrate that electrical energy is generated, the units can be connected to an included micromotor with propeller. Detailed instructions for use included. The kit contains i.a. the following components: Micro motor, 60 mV / 30 mA, Solar cell, 0.5 V / 300 mA, Flashlight, Electrolysis bath, Copper and zinc electrodes, Paper strips, Citric acid, Plastic tube with two rings, Rod magnet, Copper and constant wire.
Storage box dimensions: 300 × 170 × 40 mm