Order Entry
Sweden
ContactUsLinkComponent
Basic Gas Chromatography
Basic Gas Chromatography
Catalog # 766-0182
Supplier:  WILEY JOHN SONS
undefined
Basic Gas Chromatography
Catalog # 766-0182
Supplier:  WILEY JOHN SONS
Specifications
  • Description:
    Basic gas chromatography
  • Pk:
    1
Specifications
About this item
Basic book on gas chromatography. Principles, calculations in qualitative and quantitative analyzes. How different detectors such as TCD, FID etc. works. Structure and properties of different stationary phases and support materials. Suitable for you who have or are considering obtaining our school chromatograph GC82 12.