D-(+)-Proline
Catalog # MOLEM51773021
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:115.13 g/mol
Melting Pt:223 °C
EINECS:206-452-7
CAS Number:344-25-2
Formula:C₅H₉NO₂
MDL Number:MFCD00064317