D-(+)-Proline ≥99%, white powder
Supplier: MP Biomedicals
Certificates
About this item
CAS Number:344-25-2
:
Formula:C₅H₉NO₂
MDL Number:MFCD00064317
:115.13 g/mol
Melting Pt:225…235 °C
EINECS:206-452-7
Specification Test Results
- Optical Rotation:+84,5° to +86,5° (c=4, H2O)
- Purity:≥99%
- FTIR:Conforms to standard