Order Entry
Sweden
ContactUsLinkComponent
Pesticide reference standard, Chlorobenzene 100 µg/ml in methanol, Dr Ehrenstorfer
Pesticide reference standard, Chlorobenzene 100 µg/ml in methanol, Dr Ehrenstorfer
Catalog # EHERXA11380000ME
Supplier:  EHRENSTORFER
CAS Number:  
Pesticide reference standard, Chlorobenzene 100 µg/ml in methanol, Dr Ehrenstorfer
Catalog # EHERXA11380000ME
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 mL
  • CAS No.:
    108-90-7
  • Description:
    Chlorobenzene 100 µg/ml in methanol
  • Formula:
    C₆H₅Cl
  • Description:
    Chlorobenzene

Specifications

About this item