Saponin
Glass bottle for solids, 500 g
Katalog # MOLE50681330-500G
Marketsource
Leverantör: Molekula
Gå till:
Product Details & Documents
- CAS-nummer:8047-15-2
- MDL-nummer:MFCD00081981
- Densitet:1.015…1.02 g/cm³ (20 °C)
- Förvaringstemperatur:Ambient
- EINECS:232-462-6
Specifikationer
Specifikationer
- Pack type:Glass bottle for solids
- Pk:500 g
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



