Order Entry
Sweden
ContactUsLinkComponent
Saponin, Millipore®
Saponin, Millipore®
Catalog # 558255-100
CAS Number:  
Saponin, Millipore®
Catalog # 558255-100
CAS Number:  

Specifications

  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
Density:1.015…1.02 g/cm³ (20 °C)
EINECS:232-462-6
 :GHS07
Storage Temperature:Ambient
CAS Number:8047-15-2
MDL Number:MFCD00081981