Order Entry
Sweden
ContactUsLinkComponent
Saponin (from Quillaja Bark)
Saponin (from Quillaja Bark)
Katalog # A4518.0100
Leverantör:  PanReac AppliChem
CAS-nummer:  
Saponin (from Quillaja Bark)
Katalog # A4518.0100
Leverantör:  PanReac AppliChem
CAS-nummer:  

Specifikationer

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g
  • Seq:
    3401

Specifikationer

Om denna produkt

 :
Merck Index:13,08442
EINECS:232-462-6
CAS-nummer:8047-15-2
MDL-nummer:MFCD00081981