0,00 kr
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Sweden
ContactUsLinkComponent
Tide Fluor™ 8WS azide
Tide Fluor™ 8WS azide, Tide Fluor™ fluorescent probe
Tide Fluor™ 8WS azide, Tide Fluor™ fluorescent probe
AATB2306
:
AAT BIOQUEST
:
Tide Fluor™ 8WS azide, Tide Fluor™ fluorescent probe
AATB2306
:
AAT BIOQUEST
:
Förpackningstyp:
Plastic bottle
Förp.:
1 mg
Seq:
2070
:
:
1125.19 g/mol
:
5,II
:
GHS07