Order Entry
Sweden
ContactUsLinkComponent
TSQ zinc indicator
Irritant
TSQ zinc indicator
Catalog # AATB21254
Supplier:  AAT Bioquest
CAS Number:  
undefined
Irritant
TSQ zinc indicator
Catalog # AATB21254
Supplier:  AAT Bioquest
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    5 mg
  • Seq:
    2210

Specifications

About this item

 :
 :328.38 g/mol
ADR:5,II
 :GHS07
CAS Number:109628-27-5
Formula:C₁₇H₁₆N₂O₃S