
Pluronic® F-127 10% in water
Catalog # AATB20053
Supplier: AAT Bioquest
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:10 mL
- Seq:2275
Specifications
About this item
:
ADR:5,II
:GHS07
Storage Temperature:Ambient
CAS Number:9003-11-6
Formula:HO(C₂H₄O)a(C₃H₆O)b(C₂H₄O)aH
MDL Number:MFCD00082049