0,00 kr
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Sweden
ContactUsLinkComponent
Cyanine 5-dATP [Cy5-dATP]
Cyanine 5-dATP [Cy5-dATP]
Cyanine 5-dATP [Cy5-dATP]
Irritant
Cyanine 5-dATP [Cy5-dATP]
Catalog # AATB17038
Supplier:
AAT Bioquest
CAS Number:
Expand image
Irritant
Cyanine 5-dATP [Cy5-dATP]
Catalog # AATB17038
Supplier:
AAT Bioquest
CAS Number:
Specifications
Pack type:
Plastic bottle
Pk:
25 Nmo
Seq:
0311
Specifications
About this item
Product Details
:
:
1182.05 g/mol
ADR:
5,II
:
GHS07
Product Details