Anti-IgG Goat Polyclonal Antibody [clone: IgG] (trFluor™ Eu)
Supplier: AAT BIOQUEST
Certificates
About this item
Many biological compounds present in cells, serum or other biological fluids are naturally fluorescent, and thus the use of conventional, prompt fluorophores leads to serious limitations in assay sensitivity due to the high background caused by the autofluorescence of the biological molecules to be assayed.
- Large stokes shift
- Improved stability
- High emission yield
- Long emission half-life
Caution: For research use only.
Type: Secondary
Antigen: Immunoglobulin G
Clonality: Polyclonal
Clone: IgG
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Goat
Isotype: IgG (H, L)
Reactivity: Mouse
Specifications
- Catalog No:
- AATB16791
- AATB16598
- Antigen symbol:
- IgG
- IgG
- Antigen name:
- Immunoglobulin G
- Immunoglobulin G
- Antibody type:
- Secondary
- Secondary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- trFluor™ Eu
- trFluor™ Eu
- Clone:
- IgG
- IgG
- Reactivity:
- Mouse
- Mouse
- Host:
- Goat
- Goat
- Isotype:
- IgG (H, L)
- IgG (H, L)
- Cross adsorption:
- Min × Human
- Min × Human