Anti-IgG Donkey Polyclonal Antibody [clone: IgG] (trFluor™ Eu)
AATB62005
- Antibody type:Secondary
- Antigen name:Immunoglobulin G
- Antigen symbol:IgG
- Clonality:Polyclonal
- Clone:IgG
- Conjugation:trFluor™ Eu
- Host:Donkey
- Isotype:IgG (H, L)
- Reactivity:Rat
- Cross adsorption:Min × Human
- Pack type:Vial
- Pk:50 µG
Many biological compounds present in cells, serum or other biological fluids are naturally fluorescent, and thus the use of conventional, prompt fluorophores leads to serious limitations in assay sensitivity due to the high background caused by the autofluorescence of the biological molecules to be assayed.
- High stability and emission yield
- Low background signal
- Large stokes shift
- Long emission half-life
- Multiple applications
: For research use only.
Type: Secondary
Antigen: Immunoglobulin G
Clonality: Polyclonal
Clone: IgG
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Goat
Isotype: IgG (H, L)
Reactivity: Mouse