Order Entry
Sweden
ContactUsLinkComponent
Native Streptomyces avidinii Streptavidin (from Streptomyces avidinii), iFluor® 633
Native Streptomyces avidinii Streptavidin (from Streptomyces avidinii), iFluor® 633
Supplier:  AAT Bioquest
Certificates
undefined
Native Streptomyces avidinii Streptavidin (from Streptomyces avidinii), iFluor® 633
Supplier:  AAT Bioquest

Specifications

  • Conjugation:
    iFluor® 633
  • Protein/peptide type:
    Native
  • Source:
    Streptomyces avidinii
  • Species:
    Streptomyces avidinii
  • Protein synonyms:
    Avidin|Streptomyces avidinii

Specifications

Frequently Bought Together