Native Megathura crenulata Keyhole limpet hemocyanin (from Serum), Sulfathiazol
Catalog # AATB50030
Supplier: AAT Bioquest
CAS Number:
Specifications
- Pack type:Vial
- Pk:1 mg
- Conjugation:Sulfathiazol
- Protein/peptide type:Native
- Source:Serum
- Species:Megathura crenulata
- Protein synonyms:KLH
- Protein/peptide name:Keyhole limpet hemocyanin
Specifications
About this item
Sulfathiazol-KLH conjugate is used for developing anti-sulfathiazol antibodies that are typically used in an ELISA assay.
- High immunogenicity
- Useful for antibody development
- ELISA assay building block
Caution: For research use only.