Order Entry
Sweden
ContactUsLinkComponent
Native Streptomyces avidinii Streptavidin (from Streptomyces avidinii), Alexa Fluor® 350
Native Streptomyces avidinii Streptavidin (from Streptomyces avidinii), Alexa Fluor® 350
Catalog # AATB16890
Supplier:  AAT Bioquest
CAS Number:  
undefined
Native Streptomyces avidinii Streptavidin (from Streptomyces avidinii), Alexa Fluor® 350
Catalog # AATB16890
Supplier:  AAT Bioquest
CAS Number:  

Specifications

  • Pack type:
    Vial
  • Pk:
    1 mg
  • Conjugation:
    Alexa Fluor® 350
  • Protein/peptide type:
    Native
  • Source:
    Streptomyces avidinii
  • Species:
    Streptomyces avidinii
  • Protein synonyms:
    Avidin, Streptomyces avidinii
  • Protein/peptide name:
    Streptavidin

Specifications

About this item

XFD350 is manufactured by AAT Bioquest, and it has the same chemical structure of Alexa Fluor® 350 (Alexa Fluor® is the trademark of ThermoFisher).

  • Extremely high biotin-binding affinity
  • High quality conjugate
Caution: For research use only.