Order Entry
Sweden
ContactUsLinkComponent
Bucculite™ XdU Cell Proliferation Fluorescence Imaging Kit
  AATB22327
 :  
undefined
Bucculite™ XdU Cell Proliferation Fluorescence Imaging Kit
  AATB22327
 :  
  • Pk:
    200 Tests
Provides an alternative to anti-BrdU antibody-based assay and EdU click chemistry-based assay.
  • Alternative to anti-BrdU
  • Extremely sensitive
  • Red fluorescence
The resulted iFluor® 555-labeled DNA formed in cells is imaged with a Cy3 filter set..
Uses XdU which is incorporated into cellular DNA during DNA synthesis.