Order Entry
Sweden
ContactUsLinkComponent
Tritirachium album proteinase K
Tritirachium album proteinase K
Catalog # ROCKMB-112-0100
CAS Number:  
Tritirachium album proteinase K
Catalog # ROCKMB-112-0100
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 mg
  • CAS No.:
    39450-01-6
  • Species:
    Tritirachium album
  • Storage conditions:
    Freezer
  • Enzyme name:
    Proteinase K

Specifications

About this item