Native Streptavidin (from Streptomyces avidinii), AP (Alkaline Phosphatase)
Catalog # AATB16921
Supplier: AAT Bioquest
CAS Number:
Specifications
- Pack type:Vial
- Pk:1 mg
- Conjugation:AP (Alkaline Phosphatase)
- Protein/peptide type:Native
- Source:Streptomyces avidinii
- Protein synonyms:Avidin|Streptomyces avidinii
- Protein/peptide name:Streptavidin
Specifications
About this item
Streptavidin conjugates are widely used together with a conjugate of biotin for specific detection of a variety of proteins, protein motifs, nucleic acids and other molecules since streptavidin has a very high binding affinity for biotin.
- Extremely useful conjugate
- Multiple applications
- In-house quality control
Caution: For research use only.