Order Entry
Sweden
ContactUsLinkComponent
 
Anti-IgG Goat Polyclonal Antibody [clone: IgG] (trFluor™ Eu)
 
undefined
Anti-IgG Goat Polyclonal Antibody [clone: IgG] (trFluor™ Eu)

 

  • Catalog No:
  • AATB16755
  • AATB16518
  • Antigen symbol:
  • IgG
  • IgG
  • Antigen name:
  • Immunoglobulin G
  • Immunoglobulin G
  • Antibody type:
  • Secondary
  • Secondary
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Conjugation:
  • trFluor™ Eu
  • trFluor™ Eu
  • Clone:
  • IgG
  • IgG
  • Reactivity:
  • Mouse
  • Mouse
  • Host:
  • Goat
  • Goat
  • Isotype:
  • IgG (H, L)
  • IgG (H, L)
  • Cross adsorption: