ReadiUse™ Bio-Gel P-6 Spin Column (5)
Catalog # AATB60500
Supplier: AAT Bioquest
Specifications
- Column Format:Capillary
- Ø int.:12,00 mm
- Capacity:2 ml
- Description:ReadiUse™ bio-gel P-6 spin column
- Length:75 mm
- Pk:5
- Sample volume:30 - 100 µl ml
Specifications
About this item
ReadiUse™ Spin chromatography columns are packed with polyacrylamide P-6-DG matrices for buffer exchange and desalting applications.
- Exclusion limit of MW 6000 retained
- Exclusion limit above MW 15000 eluted
- swinging bucket centrifuge compatible
- CAS: 25034-58-6
Caution: For research use only.