Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Tide Fluor™ 3WS amine
Tide Fluor™ 3WS amine, Tide Fluor™ fluorescent probe
Tide Fluor™ 3WS amine, Tide Fluor™ fluorescent probe
Irritant
Tide Fluor™ 3WS amine, Tide Fluor™ fluorescent probe
Catalog # AATB2347
Supplier:
AAT Bioquest
CAS Number:
Expand image
Irritant
Tide Fluor™ 3WS amine, Tide Fluor™ fluorescent probe
Catalog # AATB2347
Supplier:
AAT Bioquest
CAS Number:
Specifications
Förpackningstyp:
Plastic bottle
Förp.:
1 mg
Seq:
2070
Specifications
About this item
Product Details
:
:
996 g/mol
ADR:
5,II
:
GHS07
Product Details