0,00 kr
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Sweden
ContactUsLinkComponent
Tide Fluor™ 4 acid
Tide Fluor™ 4 acid, Tide Fluor™ fluorescent probe
Irritant
Tide Fluor™ 4 acid, Tide Fluor™ fluorescent probe
Catalog # AATB2285
Supplier:
AAT BIOQUEST
CAS Number:
Expand image
Irritant
Tide Fluor™ 4 acid, Tide Fluor™ fluorescent probe
Catalog # AATB2285
Supplier:
AAT BIOQUEST
CAS Number:
Specifications
Pack type:
Plastic bottle
Pk:
10 mg
Seq:
2275
Specifications
About this item
Product Details
:
ADR:
5,II
:
562.65 g/mol
:
GHS07
Product Details