
Tide Fluor™ 5WS maleimide, Tide Fluor™ fluorescent probe
Catalog # AATB2280
Supplier: AAT Bioquest
CAS Number:
Specifications
- Förpackningstyp:Plastic bottle
- Förp.:1 mg
- Seq:2070
Specifications
About this item
:
:987.05 g/mol
ADR:5,II
:GHS07