Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Tide Fluor™ 5WS acid
Tide Fluor™ 5WS acid, Tide Fluor™ fluorescent probe
Tide Fluor™ 5WS acid, Tide Fluor™ fluorescent probe
Irritant
Tide Fluor™ 5WS acid, Tide Fluor™ fluorescent probe
Catalog # AATB2278
Supplier:
AAT Bioquest
CAS Number:
Expand image
Irritant
Tide Fluor™ 5WS acid, Tide Fluor™ fluorescent probe
Catalog # AATB2278
Supplier:
AAT Bioquest
CAS Number:
Specifications
Pack type:
Plastic bottle
Pk:
10 mg
Seq:
2275
Specifications
About this item
Product Details
:
:
864.92 g/mol
ADR:
5,II
:
GHS07
Product Details