Order Entry
Sweden
ContactUsLinkComponent
Tide Fluor™ 3 amine, Tide Fluor™ fluorescent probe
Irritant
Tide Fluor™ 3 amine, Tide Fluor™ fluorescent probe
Catalog # AATB2269
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
Irritant
Tide Fluor™ 3 amine, Tide Fluor™ fluorescent probe
Catalog # AATB2269
Supplier:  AAT BIOQUEST
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 mg
  • Seq:
    2070

Specifications

About this item

 :
 :856.69 g/mol
ADR:5,II
 :GHS07