Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Tide Fluor™ 1 acid
Tide Fluor™ 1 acid, Tide Fluor™ fluorescent probe
Tide Fluor™ 1 acid, Tide Fluor™ fluorescent probe
Tide Fluor™ 1 acid, Tide Fluor™ fluorescent probe
AATB2238
:
AAT BIOQUEST
:
Tide Fluor™ 1 acid, Tide Fluor™ fluorescent probe
AATB2238
:
AAT BIOQUEST
:
Pack type:
Plastic bottle
Pk:
100 mg
Seq:
2440
:
:
5,II
:
233.22 g/mol
:
GHS07