Order Entry
Sweden
ContactUsLinkComponent
Red 600, Protonex™
Irritant
Red 600, Protonex™
Catalog # AATB21207
Supplier:  AAT Bioquest
CAS Number:  
undefined
Irritant
Red 600, Protonex™
Catalog # AATB21207
Supplier:  AAT Bioquest
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 mg
  • Seq:
    2070

Specifications

About this item

 :
 :698.94 g/mol
ADR:5,II
 :GHS07