Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Cyanine 7 alkyne
Cyanine 7 alkyne ≥90% [equivalent to Cy7® alkyne]
Cyanine 7 alkyne ≥90% [equivalent to Cy7® alkyne]
Cyanine 7 alkyne ≥90% [equivalent to Cy7® alkyne]
Catalog # AATB164
Supplier:
AAT Bioquest
CAS Number:
Expand image
Cyanine 7 alkyne ≥90% [equivalent to Cy7® alkyne]
Catalog # AATB164
Supplier:
AAT Bioquest
CAS Number:
Specifications
Pk:
1 mg
Seq:
2070
Specifications
About this item
Product Details
:
:
833.93 g/mol
Product Details