Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
FAM-cGMP PDE
FAM-cGMP PDE substrate "Green Fluorescence"
FAM-cGMP PDE substrate "Green Fluorescence"
Irritant
FAM-cGMP PDE substrate "Green Fluorescence"
Catalog # AATB13604
Supplier:
AAT BIOQUEST
CAS Number:
Expand image
Irritant
FAM-cGMP PDE substrate "Green Fluorescence"
Catalog # AATB13604
Supplier:
AAT BIOQUEST
CAS Number:
Specifications
Pack type:
Plastic bottle
Pk:
0,5 µmol
Seq:
9030
Specifications
About this item
Product Details
:
ADR:
5,II
:
GHS07
Product Details