Order Entry
Sweden
ContactUsLinkComponent
Mca-PLAQAV-Dpa-RSSSR-NH2
Irritant
Mca-PLAQAV-Dpa-RSSSR-NH2
Catalog # AATB13560
Supplier:  AAT Bioquest
CAS Number:  
undefined
Irritant
Mca-PLAQAV-Dpa-RSSSR-NH2
Catalog # AATB13560
Supplier:  AAT Bioquest
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 mg
  • Seq:
    2070

Specifications

About this item

 :
ADR:5,II
 :GHS07