Table optics, TESS OE1
Katalog # 765-1317
Leverantör: PHYWE
Specifikationer
- Description:TESS Optics OE1
- Pk:1
Specifikationer
Om denna produkt
PHYWE table optics for laboratory work with mirrors, light refraction, lenses, eye function and light refraction. Content:
- 1 light box with halogen lamp 12 V/20 W
- 2 columns
- 6 lens models
- 1 cuvette, double, semicircular
- 1 mirror, plan
- 1 mirror, concave-convex
- 1 optical disc
- 1 storage box
- Delivered in plywood box