Table optics, TESS OE2
Katalog # 765-1318
Leverantör: PHYWE
Specifikationer
- Description:TESS Optics OE2
- Pk:1
Specifikationer
Om denna produkt
The kit contains 30 components i.a. an optical bench, lenses in holders, grids, polarization filters. Together with TESS 1, a total of 70 different experiments can be performed, e.g. study of telescopes, microscopes, the human eye and the plane of polarization.
- Delivered in a plywood box