Green laser light source
Laser light source, green
Catalog # 765-1007
Supplier: KVANT LIFE SCIENCE
Jump to:
Product Details & Documents
Light box with five parallel-mounted laser diodes (wavelength 635 nm). The diodes are equipped with a collimator and a 5 mm cylindrical lens, which means that the beam path can be followed before, through and after the lens as five sharp and clearly visible, parallel light beams.
Specifications
- Description:Laser light source, green