Om denna produkt
Sani-Tech® Ultra-65 represent the next generation in ultra-pure platinum silicone tubing with ultra-low extractable and longer pump life. These tubings have been designed by our Saint-Gobain Life Sciences R&D team in order to fully comply with the stringent requirements of the biopharmaceutical market. Sani-Tech® Ultra-65 come with laser marked part numbers and lot codes to provide total product traceability.
- Two hardnesses available, 50 and 65 shore
- Biopharmaceutical grade
- Ultra-low extractables
- Excellent flexibility
- Laser etched for lot traceability
- Superior pump life
- Temperature range from −62 to +260 °C
- Validation guide summary
- BPOG extractable report
Fluid integrity is vitally important in many biopharmaceutical applications. The manufacturing process is carefully controlled from receiving through production. In all our multiple production sites (US, France and India) our tubing is produced and packaged in double bags within an ISO 7 clean room. Ultra-low TOCs ensure that contamination by extractables is kept to an absolute minimum.
Typical applications: Sterile fill and transfers, drug delivery, media/buffer processing, pumping, cell culture and harvest, chromatography, final fill, filtration and fermentation