Didactic laser 1, red
Didactic Laser 1, red
Catalog # 765-1012
Supplier: KVANT LIFE SCIENCE
Jump to:
Product Details & Documents
Laser type: Diode, Wavelength: 635 nm.
Power: 1mW / Class II
Dimensions: 35 × 300 mm
Specifications
- Description:Didactic Laser 1, red
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



