DURAN® Glass Capillaries
Supplier: Schott AG
Certificates
About this item
High precision capillaries made from borosilicate 3.3 glass.
- High chemical resistance
- High hydrolytical resistance
- High temerature resistance
- High resistance to thermal shock