Order Entry
Sweden
ContactUsLinkComponent
Recombinant Mouse PECAM1 (from HEK293 cells), His-tag
Recombinant Mouse PECAM1 (from HEK293 cells), His-tag
  ABNOP9020.10UG
 :  Abnova
 :  
Recombinant Mouse PECAM1 (from HEK293 cells), His-tag
  ABNOP9020.10UG
 :  Abnova
 :  

 

  • Pk:
    10 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Mouse
  • Tag sequence:
    His-tag
  • Gene ID:
    Q08481
  • Protein/peptide name:
    PECAM1
  • Sequence:
    18a.a.-590a.a.