Recombinant Mouse SPP1 (from HEK293 cells), His-tag
Catalog # ABNOP9189.20UG
Supplier: Abnova
CAS Number:
Specifications
- Pk:20 µG
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Mouse
- Tag sequence:His-tag
- Protein/peptide name:SPP1