Order Entry
Sweden
ContactUsLinkComponent
 
Recombinant Mouse UCHL3 (from Baculovirus), His-tag
  ABNOP7981.50UG
 :  Abnova
 :  
Recombinant Mouse UCHL3 (from Baculovirus), His-tag
  ABNOP7981.50UG
 :  Abnova
 :  

 

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    Baculovirus
  • Species:
    Mouse
  • Tag sequence:
    His-tag
  • Gene ID:
    Q9JKB1
  • Protein/peptide name:
    UCHL3
  • Sequence:
    1a.a.-230a.a.