Tygon® F-4040-A tubing resists the swelling and hardening caused by hydrocarbon-based fluids - significantly reducing the risk of failure due to cracks and leaks. Its minimum extractability safeguards the liquid or vapour being transferred against adulteration. It is extremely flexible and ideal for small engines due to its ability to maneuver tight places and simplify installation.
- Resists embrittlement
- Compatible with most hydrocarbons
- Resistant to swelling
- Highly flexible, easy to install
- Specifically developed for fuels and lubricants
- Ozone and UV light resistant
This product’s translucent yellow colour stands out for identification purposes yet allows you to easily view and monitor the fuels and lubricants flowing through it. A consistent performer lot after lot, Tygon® Fuel and Lubricant Tubing is the most requested fuel and lubricant tubing for a variety of applications - from small engine fuel lines to coolant transfer. It is ideal for applications such as small engine fuel lines, general automotive, recreational vehicles, lawn and garden equipment, coolant transfer, heating fuels, cutting compounds, polishing equipment and lubrication lines.
Hardness: 57 (Shore A)
- Catalog No:
- VERNAAG00165
- VERNAAG00032
- VERNAAG00700
- VERNAAG00053
- VERNAAG00046
- VERNAAG00036
- VERNAAG00038
- VERNAAG00007
- VERNAAG00012
- VERNAAG00017
- VERNAAG00029
- VERNAAG00027
- Max. temperature:
- 74 °C
- 74 °C
- 74 °C
- 74 °C
- 74 °C
- 74 °C
- 74 °C
- 74 °C
- 74 °C
- 74 °C
- 74 °C
- 74 °C
- Min. temperature:
- −37 °C
- −37 °C
- −37 °C
- −37 °C
- −37 °C
- −37 °C
- −37 °C
- −37 °C
- −37 °C
- −37 °C
- −37 °C
- −37 °C