Anti-Caspase-3 Rabbit Recombinant Antibody [clone: R04-5C8]
Catalog # ABNORAB01537.100UL
Supplier: Abnova
Specifications
- Antigen symbol:Caspase-3
- Clonality:Recombinant
- Clone:R04-5C8
- Host:Rabbit
- Reactivity:Human
- Immunogen:Recombinant rabbit monoclonal antibody raised against synthetic peptide of human Caspase-3.
- Pk:100 µl