Clamps, Zinc Die Cast
These clamps are zinc diecasted, nickel plated with vinyl coated jaws. Rods are 10 mm in diameter.
Advantages of zinc diecasting:
Better energy efficiency during casting process thanks to a lower melting point, when compared to aluminum or malleable cast iron
High recycling proportion of at least 75% of raw material used
High stability also of thin-walled parts e.g. bossheads
Excellent cost-performance ratio of our basic assortment
Good corrosion resistance - protection through natural passivation (reaction of zinc with oxygen at the surface)
Corrosion resistance can be further enhanced by powder coating or nickel plating