Order Entry
Sweden
ContactUsLinkComponent
Experimentation kits, ion exchange chromatography
Experimentation kits, ion exchange chromatography
  EDVO243
 :  EDVOTEK
undefined
Experimentation kits, ion exchange chromatography
  EDVO243
 :  EDVOTEK
  • Description:
    Ion Exchange Chromatography
  • Pk:
    1
Most molecules have a net charge within a pH range of 2 to 10. When the pH is altered, the net charge on molecules can change drastically. In this experiment, a mixture of two chemicals is absorbed onto a solid support ion-exchange column and separated during elution under conditions that influence their net charge.
  • For 6 lab groups
  • Requires 60 to 90 minutes
Also required: Spectrophotometer and cuvettes, ring stands and clamps, and 5 ml pipettes.
 : Kit includes instructions, ion exchanger, chemical mixture, potassium acetate buffer, and chromatography columns.