Order Entry
Sweden
ContactUsLinkComponent
Kits: in search of my father, Ready-to-Load™
Kits: in search of my father, Ready-to-Load™
Catalog # EDVOS-49
Supplier:  EDVOTEK
undefined
Kits: in search of my father, Ready-to-Load™
Catalog # EDVOS-49
Supplier:  EDVOTEK
Specifications
  • Description:
    In Search of My Father
  • Pk:
    1
Specifications
About this item
Solve the mystery of two boys separated from their parents a decade ago. Their biological mother is identified by mitochondrial DNA and their biological father from chromosomal DNA.
  • For 10 lab groups
  • Complete in 45 minutes
Also required: Electrophoresis tank and power supply.
Delivery information: Kit includes instructions, Ready-to-Load™ dye samples, agarose powder, practice gel loading solution, electrophoresis buffer, and microtipped transfer pipettes.