Data logger, USBLink
Data logger, USBlink, with a eight input sensor interface
Catalog # 620-1795
Supplier: FOURIER SYSTEMS
Jump to:
Product Details & Documents
The USBLink is a sensor interface, converting any computer into a science data logging learning station. The USBLink simple 'plug-and-play' unit is highly functional, featuring four-input interface, with an option for up to eight simultaneously connected sensors via USB to the computer. The unit offers automatic sensor recognition, fast sampling rate and sample streaming of up to 10,000 samples per second.
- Simple to operate
- Simulataneous data collection for up to eight sensors
- Works with full range of Fourier sensors
- Powered directly by any PC USB port
- Incorporates MultiLab data analysis software
Specifications
- Description:Data logger, USBlink, with a eight input sensor interface
- Cat. No.:620-1795
- Interface:USB