Blue Super-Duty Autoclavable SMS Muff Covers
Supplier: KEYSTONE CLEANROOM PRODUCTS
Certificates
About this item
This 88 g/m² SMS autoclavable muff cover provides an excellent microbial barrier that is ideal for pharmaceutical production applications.
- Eliminates autoclave tape residue
- Hydrophobic, tear-resistant, and puncture-resistant
- Creates secondary level of protection during aseptic presentation and setup of equipment
- Designed for autoclave cycles (121 to 135 °C)
- Manufactured and packed in a certified ISO 8 cleanroom
- Used in conjunction with a secondary autoclave bag, it creates an effective wrapping system
- Made in USA
Eliminate the need for cutting sheets of sterilisation wrapping materials and securing them with autoclave tape and rubber bands. Cost-effective, time-saving approach standardises the equipment preparation process, minimises particulate generation, while also ensuring good aseptic technique upon removal after sterilisation.
Certifications: Certificates of conformance are sent with every shipment.
Packaging: Cleanroom packed in sealed, labelled poly-bags with the lot number and date of manufacture. Covers are packed 100 per bag.